Få mere at vide
1-Bromopropane-d7, TRC
Artikelnummer. 30333087
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30333087 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30333087 Leverandør Toronto Research Chemicals Leverandørnr. TRCB687138500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopropane-d7
CAS 61909-26-0
Molekylær formel C3 D7 Br
Mængde 500 mg
Synonym 1-Propyl Bromide-d7, 3-Bromopropane-d7, Abzol JG-d7, Ascusol MC-d7, Drysolv-d7, Leksol-d7, Propyl Bromide-d7, n-Propyl Bromide-d7, 3-Bromopropane-1,1,1,2,2,3,3-d7, Heptadeuteriopropyl Bromide, Propyl-d7 Bromide
InChI formel InChI=1S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])Br
IUPAC navn 1-bromo-1,1,2,2,3,3,3-heptadeuteriopropane
Molekylvægt (g/mol) 130.03
Formel vægt 129.0171