Få mere at vide
1-Bromopropane-2,2,3,3,3-d5, TRC
Artikelnummer. 30346106
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30346106 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346106 Leverandør Toronto Research Chemicals Leverandørnr. TRCB687139100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopropane-2,2,3,3,3-d5
CAS 61066-67-9
Molekylær formel C3 D5 H2 Br
Mængde 100 mg
Synonym Propane-1,1,1,2,2-d5, 3-bromo- (9CI), 1-Bromopropane-2,2,3,3,3-d5
InChI formel InChI=1S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])CBr
IUPAC navn 3-bromo-1,1,1,2,2-pentadeuteriopropane
Molekylvægt (g/mol) 128.02
Formel vægt 127.0045