Få mere at vide
1-Bromopropane-2,2,3,3,3-d5, CDN
Artikelnummer. 30323490
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323490 1 g -
30313576 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323490 Leverandør CDN Leverandørnr. CDND02251G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopropane-2,2,3,3,3-d5
CAS 61066-67-9
Molekylær formel C3 D5 H2 Br
Mængde 1 g
Synonym Propane-1,1,1,2,2-d5, 3-bromo- (9 CI), 1-Bromopropane-2,2,3,3,3-d5
InChI formel InChI=1 S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CBr
IUPAC navn 3-bromo-1,1,1,2,2-pentadeuteriopropane
Molekylvægt (g/mol) 128.02
Formel vægt 127.0045 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity