Få mere at vide
1-Bromopropane-1,1,3,3,3-d5, CDN
Artikelnummer. 30315138
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315138 0.5 g 0.5g
30321742 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315138 Leverandør CDN Leverandørnr. CDND03430.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopropane-1,1,3,3,3-D5
CAS 163400-20-2
Molekylær formel C3 D5 H2 Br
Mængde 0.5 g
Synonym Propane-1,1,1,3,3-d5, 3-bromo- (9 CI), 1-Bromopropane-1,1,3,3,3-d5, 3-Bromopropane-1,1,1,3,3-d5, 1-Bromopropane-d5, 1-Bromopropane-1,1,3,3,3-D5
InChI formel InChI=1 S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3,3D2
SMIL [2 H]C([2 H])([2 H])CC([2 H])([2 H])Br
IUPAC navn 1-bromo-1,1,3,3,3-pentadeuteriopropane
Molekylvægt (g/mol) 128.0225
Formel vægt 127.0045 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity