missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoperfluorononane, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-Bromoperfluorononane is a specialty product for proteomics research applications. It is also used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkali metals and alkaline earth metals.
1-Bromoperfluorononane is a specialty product for proteomics research applications. It is also used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkali metals and alkaline earth metals.
Specifications
Melting Point | 27°C to 30°C |
Density | 1.9 |
Boiling Point | 159°C |
Flash Point | None |
Linear Formula | CF3(CF2)8Br |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 548.97 |
Percent Purity | 98% |
Chemical Name or Material | 1-Bromoperfluorononane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'