Få mere at vide
1-Bromopentane-4,4,5,5,5-d5, CDN
Artikelnummer. 30327192
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327192 1 g -
30326601 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327192 Leverandør CDN Leverandørnr. CDND66291G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopentane-4,4,5,5,5-D5
CAS 83418-34-2
Molekylær formel C5 D5 H6 Br
Mængde 1 g
Synonym Pentane-1,1,1,2,2-d5, 5-bromo- (9 CI), 1-Bromopentane-4,4,5,5,5-d5, 5-Bromopentane-1,1,1,2,2-d5, 1-Bromo-4,4,5,5,5-pentadeuteriopentane, 1-Bromopentane-d5, 1-Bromopentane-4,4,5,5,5-D5
InChI formel InChI=1 S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCBr
IUPAC navn 5-bromo-1,1,1,2,2-pentadeuteriopentane
Molekylvægt (g/mol) 156.0757
Formel vægt 155.0358 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity