Få mere at vide
1-Bromopentane-1-d1, CDN
Artikelnummer. 30319560
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319560 0.05 g -
30319105 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319560 Leverandør CDN Leverandørnr. CDND69320.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopentane-1-D
CAS 148404-92-6
Molekylær formel C5 D H10 Br
Mængde 0.05 g
Synonym Pentane-1-d, 1-bromo- (9 CI), 1-Bromopentane-1-d, 1-Bromopentane-1-d1, 1-Bromopentane-d, 1-Bromopentane-1-D
InChI formel InChI=1 S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i5D
SMIL [2 H]C(Br)CCCC
IUPAC navn 1-bromo-1-deuteriopentane
Molekylvægt (g/mol) 152.051
Formel vægt 151.0107 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity