Få mere at vide
1-Bromopentadecane-1,1,2,2-d4, CDN
Artikelnummer. 30312836
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312836 0.25 g -
30317786 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312836 Leverandør CDN Leverandørnr. CDND64590.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromopentadecane-1,1,2,2-D4
CAS 1219798-87-4
Molekylær formel C15 D4 H27 Br
Mængde 0.25 g
Synonym 1-Bromopentadecane-1,1,2,2-d4
InChI formel InChI=1 S/C15H31Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h2-15H2,1H3/i14D2,15D2
SMIL [2 H]C([2 H])(Br)C([2 H])([2 H])CCCCCCCCCCCCC
IUPAC navn 1-bromo-1,1,2,2-tetradeuteriopentadecane
Molekylvægt (g/mol) 295.3353
Formel vægt 294.186 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity