Få mere at vide
1-Bromooctadecane-18,18,18-d3, CDN
Artikelnummer. 30311324
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311324 0.1 g 0.1g
30313379 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311324 Leverandør CDN Leverandørnr. CDND39970.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromooctadecane-18,18,18-D3
CAS 349553-85-1
Molekylær formel C18 D3 H34 Br
Mængde 0.1 g
Synonym Octadecane-1,1,1-d3, 18-bromo- (9 CI), 1-Bromooctadecane-d3, 1-Bromooctadecane-18,18,18-d3, 1-Bromooctadecane-18,18,18-D3, 18-Bromooctadecane-1,1,1-d3, Octadecyl-18,18,18-d3 bromide, Stearyl-18,18,18-d3 bromide
InChI formel InChI=1 S/C18H37Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCCCBr
IUPAC navn 18-bromo-1,1,1-trideuteriooctadecane
Molekylvægt (g/mol) 336.4089
Formel vægt 335.2267 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity