missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)isoquinoline hydrobromide, 97%, Thermo Scientific™

Catalog Number p-2694506
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
337508-56-2
C10H9Br2N
302.997
MFCD03086096
RWSWRJTWPXSENX-UHFFFAOYSA-N
1-bromomethyl isoquinoline hydrobromide, 1-bromomethylisoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide 1:1, 1-bromomethylisoquinoline hbr, 1-bromomethyl-isoquinolin hydrobromide, 1-bromomethyl-isoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide
2776245
1-(bromomethyl)isoquinoline;hydrobromide
C1=CC=C2C(=C1)C=CN=C2CBr.Br
This item is not returnable. View return policy
337508-56-2
C10H9Br2N
302.997
MFCD03086096
RWSWRJTWPXSENX-UHFFFAOYSA-N
1-bromomethyl isoquinoline hydrobromide, 1-bromomethylisoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide 1:1, 1-bromomethylisoquinoline hbr, 1-bromomethyl-isoquinolin hydrobromide, 1-bromomethyl-isoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide
2776245
1-(bromomethyl)isoquinoline;hydrobromide
C1=CC=C2C(=C1)C=CN=C2CBr.Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 303
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)isoquinoline hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'