Få mere at vide
1-Bromohexane-4,4,5,5,6,6,6-d7, CDN
Artikelnummer. 30316335
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316335 0.25 g
30313023 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316335 Leverandør CDN Leverandørnr. CDND73320.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromohexane-4,4,5,5,6,6,6-D7
CAS 1276197-43-3
Molekylær formel C6 D7 H6 Br
Mængde 0.25 g
Synonym 6-Bromohexane-1,1,1,2,2,3,3-d7, 1-Bromohexane-4,4,5,5,6,6,6-d7, 1-Bromohexane-d7, 1-Bromohexane-4,4,5,5,6,6,6-D7
InChI formel InChI=1 S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCBr
IUPAC navn 6-bromo-1,1,1,2,2,3,3-heptadeuteriohexane
Molekylvægt (g/mol) 172.1146
Formel vægt 171.064 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity