Få mere at vide
1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-d11, CDN
Artikelnummer. 30322050
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322050 1 g -
30316083 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322050 Leverandør CDN Leverandørnr. CDND65091G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-D11
CAS 2159-17-3
Molekylær formel C6 D11 H2 Br
Mængde 1 g
Synonym Hexane-1,1,1,2,2,3,3,4,4,5,5-d11, 6-bromo- (7 CI, 9 CI), 6-Bromohexane-1,1,1,2,2,3,3,4,4,5,5-d11, 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-d11, 1-Bromohexane-d11, 1-Bromohexane-2,2,3,3,4,4,5,5,6,6,6-D11
InChI formel InChI=1 S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CBr
IUPAC navn 6-bromo-1,1,1,2,2,3,3,4,4,5,5-undecadeuteriohexane
Molekylvægt (g/mol) 176.1392
Formel vægt 175.0891 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity