Få mere at vide
1-Bromohexadecane-16,16,16-d3, CDN
Artikelnummer. 30315920
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315920 0.1 g
30310648 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315920 Leverandør CDN Leverandørnr. CDND39860.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromohexadecane-16,16,16-D3
CAS 284474-40-4
Molekylær formel C16 D3 H30 Br
Mængde 0.1 g
Synonym Hexadecane-1,1,1-d3, 16-bromo- (9 CI), 16-Bromohexadecane-1,1,1-d3, 1-Bromohexadecane-16,16,16-d3, 1-Bromohexadecane-16,16,16-D3, 1-Hexadecyl-16,16,16-d3 bromide, Cetyl-16,16,16-d3 bromide
InChI formel InChI=1 S/C16H33Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCBr
IUPAC navn 16-bromo-1,1,1-trideuteriohexadecane
Molekylvægt (g/mol) 308.3557
Formel vægt 307.1954 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity