Få mere at vide
1-Bromoheptane-d15, CDN
Artikelnummer. 30319414
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319414 0.5 g
30326774 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319414 Leverandør CDN Leverandørnr. CDND17240.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoheptane-D15
CAS 98195-42-7
Molekylær formel C7 D15 Br
Mængde 0.5 g
Synonym Heptane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-d15, 7-bromo- (9 CI), Heptyl-d15 bromide, 7-Bromoheptane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-d15, 1-Bromoheptane-d15, 1-Bromoheptane-D15
InChI formel InChI=1 S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br
IUPAC navn 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptane
Molekylvægt (g/mol) 194.1904
Formel vægt 193.1299 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity