Få mere at vide
1-Bromoheptane-7,7,7-d3, CDN
Artikelnummer. 30311945
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311945 1 g -
30311562 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311945 Leverandør CDN Leverandørnr. CDND52151G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoheptane-7,7,7-D3
CAS 344253-18-5
Molekylær formel C7 D3 H12 Br
Mængde 1 g
Synonym Heptane-1,1,1-d3, 7-bromo- (9 CI), 7-Bromoheptane-1,1,1-d3, 1-Bromoheptane-7,7,7-d3, 1-Bromoheptane-7,7,7-D3
InChI formel InChI=1 S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCBr
IUPAC navn 7-bromo-1,1,1-trideuterioheptane
Molekylvægt (g/mol) 182.1165
Formel vægt 181.0545 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity