Få mere at vide
1-Bromoheptane-6,6,7,7,7-d5, CDN
Artikelnummer. 30318803
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318803 0.5 g 0.5g
30317610 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318803 Leverandør CDN Leverandørnr. CDND57540.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoheptane-6,6,7,7,7-D5
CAS 1219805-66-9
Molekylær formel C7 D5 H10 Br
Mængde 0.5 g
Synonym 7-Bromoheptane-1,1,1,2,2-d5, 1-Bromoheptane-6,6,7,7,7-d5, 1-Bromoheptane-6,6,7,7,7-D5
InChI formel InChI=1 S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCBr
IUPAC navn 7-bromo-1,1,1,2,2-pentadeuterioheptane
Molekylvægt (g/mol) 184.1288
Formel vægt 183.0671 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity