Få mere at vide
1-Bromoeicosane-20,20,20-d3, CDN
Artikelnummer. 30318817
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318817 0.1 g
30313862 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318817 Leverandør CDN Leverandørnr. CDND52520.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromoeicosane-20,20,20-D3
CAS 202480-72-6
Molekylær formel C20 D3 H38 Br
Mængde 0.1 g
Synonym Eicosane-1,1,1-d3, 20-bromo- (9 CI), 20-Bromoeicosane-1,1,1-d3, 1-Bromoeicosane-20,20,20-d3, 1-Bromoeicosane-20,20,20-D3, Eicosyl-ω-d3 bromide
InChI formel InChI=1 S/C20H41Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h2-20H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCCCCCBr
IUPAC navn 20-bromo-1,1,1-trideuterioicosane
Molekylvægt (g/mol) 364.462
Formel vægt 363.258 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity