Få mere at vide
1-Bromodecane-9,9,10,10,10-d5, CDN
Artikelnummer. 30325964
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325964 0.1 g
30314174 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325964 Leverandør CDN Leverandørnr. CDND58900.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromodecane-9,9,10,10,10-D5
CAS 1219802-02-4
Molekylær formel C10 D5 H16 Br
Mængde 0.1 g
Synonym 1-Bromodecane-d5, 1-Bromodecane-9,9,10,10,10-D5, 10-Bromodecane-1,1,1,2,2-d5, 1-Bromodecane-9,9,10,10,10-d5, 1-Decyl-9,9,10,10,10-d5 bromide
InChI formel InChI=1 S/C10H21Br/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCBr
IUPAC navn 10-bromo-1,1,1,2,2-pentadeuteriodecane
Molekylvægt (g/mol) 226.2086
Formel vægt 225.114 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity