Få mere at vide
1-Bromobutane-d9, TRC
Artikelnummer. 30348535
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30348535 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348535 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6818335G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromobutane-d9
CAS 98195-36-9
Molekylær formel C4D9Br
Mængde 5 g
Synonym 1-Bromobutane-d9, 1-Butyl Bromide-d9, Butyl Bromide-d9, n-Butyl Bromide-d9, 4-Bromobutane-1,1,1-d9
InChI formel InChI=1S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2,4D2
SMIL BrC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1-bromobutane-1,1,2,2,3,3,4,4,4-d9
Molekylvægt (g/mol) 146.07
Formel vægt 145.05