Få mere at vide
1-Bromobutane-2,2-d2, TRC
Artikelnummer. 30347010
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30347010 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347010 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68183450MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromobutane-2,2-d2
CAS 55724-40-8
Molekylær formel C4 D2 H7 Br
Mængde 50 mg
Synonym Butane-2,2-d2, 1-bromo- (9CI), 1-Bromobutane-2,2-d2, 1-Bromobutane-2,2-D2, 1-Bromobutane-d2, n-Butyl bromide-d2
InChI formel InChI=1S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3/i3D2
SMIL [2H]C([2H])(CC)CBr
IUPAC navn 1-bromo-2,2-dideuteriobutane
Molekylvægt (g/mol) 139.0306
Formel vægt 138.0013