Få mere at vide
1-Bromobutane-2,2-d2, CDN
Artikelnummer. 30320336
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320336 1 g
30326032 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320336 Leverandør CDN Leverandørnr. CDND65721G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromobutane-2,2-d2
CAS 55724-40-8
Molekylær formel C4 D2 H7 Br
Mængde 1 g
Synonym Butane-2,2-d2, 1-bromo- (9 CI), 1-Bromobutane-2,2-d2, 1-Bromobutane-2,2-D2, 1-Bromobutane-d2, n-Butyl bromide-d2
InChI formel InChI=1 S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3/i3D2
SMIL [2 H]C([2 H])(CC)CBr
IUPAC navn 1-bromo-2,2-dideuteriobutane
Molekylvægt (g/mol) 139.0306
Formel vægt 138.0013 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity