Få mere at vide
1-Bromo-4-nitrobenzene-d4, TRC
Artikelnummer. 30331864
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30331864 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331864 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68617950MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-4-nitrobenze-d4
CAS 350820-19-8
Molekylær formel C6 2H4 Br N O2
Mængde 50 mg
Synonym Benzene-1,2,4,5-d4, 3-bromo-6-nitro- (9CI), 1-Bromo-2,3,5,6-tetradeuterio-4-nitrobenzene, 1-Bromo-4-nitrobenzene-d4, 3-Bromo-6-nitrobenzene-1,2,4,5-d4, 4-Bromonitrobenzene-d4, 1-Bromo-4-nitrobenzene-D4
InChI formel InChI=1S/C6H4BrNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Br)[N+](=O)[O-]
IUPAC navn 1-bromo-2,3,5,6-tetradeuterio-4-nitrobenzene
Molekylvægt (g/mol) 206.03
Formel vægt 204.9676