missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Bromo-4-n-phthalimido-2-butanone, TRC

Artikelnummer. 30331382
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30331382 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30331382 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69874825MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1-Bromo-4-n-phthalimido-2-butanone
CAS 51132-00-4
Molekylær formel C12 H10 Br N O3
Mængde 25 mg
InChI formel InChI=1S/C12H10BrNO3/c13-7-8(15)5-6-14-11(16)9-3-1-2-4-10(9)12(14)17/h1-4H,5-7H2
SMIL BrCC(=O)CCN1C(=O)c2ccccc2C1=O
IUPAC navn 2-(4-bromo-3-oxobutyl)isoindole-1,3-dione
Molekylvægt (g/mol) 296.117
Formel vægt 294.984
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.