Få mere at vide
1-Bromo-4-(methylsulfinyl)benzene, TRC
Artikelnummer. 30343463
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343463 250 mg
30341728 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30343463 Leverandør Toronto Research Chemicals Leverandørnr. TRCB697738250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-4-(methylsulfinyl)benzene
CAS 934-71-4
Molekylær formel C7H7BrOS
Mængde 250 mg
InChI formel InChI=1S/C7H7BrOS/c1-10(9)7-4-2-6(8)3-5-7/h2-5H,1H3
SMIL BrC1=CC=C(S(C)=O)C=C1
IUPAC navn 1-bromo-4-(methylsulfinyl)benzene
Molekylvægt (g/mol) 219.1
Formel vægt 217.94