Få mere at vide
1-Bromo-3-phenoxybenzene, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | PBDE 2 (3-Bromodiphenyl Ether) |
| CAS | 6876-00-2 |
| Molekylær formel | C12 H9 Br O |
| Mængde | 25 g |
| Synonym | Benzene, 1-bromo-3-phenoxy-, Ether, m-bromophenyl phenyl (6CI,7CI,8CI), 1-Bromo-3-phenoxybenzene, 3-Bromodiphenyl ether, 3-Bromophenyl phenyl ether, 3-Phenoxy-1-bromobenzene, 3-Phenoxybromobenzene, 3-Phenoxyphenyl bromide, BDE 2, NSC 57095, PBDE 2, m-Bromodiphenyl ether, m-Bromophenoxybenzene, m-Bromophenyl phenyl ether, m-Phenoxybromobenzene, m-Phenoxyphenyl bromide |
| InChI formel | InChI=1S/C12H9BrO/c13-10-5-4-8-12(9-10)14-11-6-2-1-3-7-11/h1-9H |
| SMIL | Brc1cccc(Oc2ccccc2)c1 |
| IUPAC navn | 1-bromo-3-phenoxybenzene |
| Molekylvægt (g/mol) | 249.1 |
| Formel vægt | 247.9837 |