Få mere at vide
1-Bromo-3-methylbutane-d7, TRC
Artikelnummer. 30335256
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30335256 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335256 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68555725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-3-methylbutane-d7
CAS 1795011-78-7
Molekylær formel C5H4D7Br
Mængde 25 mg
Synonym 3-Methyl-1-bromobutane-d7, 3-Methylbutyl-d7 Bromide, 4-Bromo-2-methylbutane-d7, Isoamyl-d7 Bromide, Isobutylmethyl-d7 Bromide, Isopentyl-d7 Bromide, NSC 9240-d7
InChI formel InChI=1S/C5H11Br/c1-5(2)3-4-6/h5H,3-4H2,1-2H3/i1D3,2D3,5D
SMIL [2H]C([2H])([2H])C([2H])(CCBr)C([2H])([2H])[2H]
IUPAC navn 4-bromo-1,1,1,2-tetradeuterio-2-(trideuteriomethyl)butane
Molekylvægt (g/mol) 158.09
Formel vægt 157.0484