Learn More
1-Bromo-3,5-dimethoxybenzene, Mikromol™
Product Code. 30049123
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049123 100 mg 100mg
30049150 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049123 Supplier Mikromol Supplier No. MM0066.09
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Bromo-3,5-dimethoxybenzene
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-3,5-dimethoxybenzene
CAS 20469-65-2
Analyte or Component Names 1-Bromo-3,5-dimethoxybenzene
Shipping Condition Room Temperature
Molecular Formula C8 H9 Br O2
Certifications/Compliance ISO 17025
Synonym 1-Bromo-3,5-dimethoxybenzene, 3,5-Dimethoxy-1-bromobenzene, 3,5-Dimethoxybromobenzene, 3,5-Dimethoxyphenyl bromide, 5-Bromo-1,3-dimethoxybenzene
InChI Formula InChI=1S/C8H9BrO2/c1-10-7-3-6(9)4-8(5-7)11-2/h3-5H,1-2H3
SMILES COc1cc(Br)cc(OC)c1
IUPAC Name 1-bromo-3,5-dimethoxybenzene
Molecular Weight (g/mol) 217.06
Quantity 100 mg
Formula Weight 215.9786 g/mol