missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,3,3-trifluoroacetone, 97%, Thermo Scientific Chemicals

Catalog Number p-7022259
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
431-35-6
C3H2BrF3O
190.95
MFCD00039237
ONZQYZKCUHFORE-UHFFFAOYSA-N
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
79008
3-bromo-1,1,1-trifluoropropan-2-one
FC(F)(F)C(=O)CBr
This item is not returnable. View return policy
431-35-6
C3H2BrF3O
190.95
MFCD00039237
ONZQYZKCUHFORE-UHFFFAOYSA-N
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
79008
3-bromo-1,1,1-trifluoropropan-2-one
FC(F)(F)C(=O)CBr

1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Melting Point <-80°C
Density 1.822
Boiling Point 85°C to 87°C
Flash Point −2°C (28°F)
Odor Pungent
Refractive Index 1.376
Quantity 5 g
UN Number UN2924
Beilstein 1703387
Solubility Information Immiscible with water.
Formula Weight 190.95
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,3,3-trifluoroacetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'