Få mere at vide
1-Bromo-2-methylpropane-d7, TRC
Artikelnummer. 30336764
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30336764 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336764 Leverandør Toronto Research Chemicals Leverandørnr. TRCB685663100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-2-methylpropane-d7
CAS 344299-41-8
Molekylær formel C4 D7 H2 Br
Mængde 100 mg
Synonym 1-Methyl-2-propyl Bromide-d7, 2-Methylpropyl Bromide-d7, Bromoisobutane-d7, Isobutyl Bromide-d7, NSC 8416-d7, iso-Butyl Bromide-d7
InChI formel InChI=1S/C4H9Br/c1-4(2)3-5/h4H,3H2,1-2H3/i1D3,2D3,4D
SMIL [2H]C([2H])([2H])C([2H])(CBr)C([2H])([2H])[2H]
IUPAC navn 2-(bromomethyl)-1,1,1,2,3,3,3-heptadeuteriopropane
Molekylvægt (g/mol) 144.06
Formel vægt 143.0327