missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%

Brand:  Alfa Aesar™ H32650.06

Additional Details : CAS Number : 286932-57-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15435397

  • 808.00 DKK / 5g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

286932-57-8
258.998
VVVFPEJORKPEST-UHFFFAOYSA-N
25067359
C1=CC(=C(C=C1OC(F)(F)F)Br)F
C7H3BrF4O
MFCD02181186
2-bromo-1-fluoro-4-trifluoromethoxy benzene, 1-bromo-2-fluoro-5-trifluoromethoxy benzene, 3-bromo-4-fluoro trifluoromethoxy benzene, 3-bromo-4-fluorotrifluoroanisole, 2-fluoro-5-trifluoromethoxy-bromobenzene, 3-bromo-4-fluorophenyl trifluoromethyl ether, 3-bromo-4-fluoro-alpha,alpha,alpha-trifluoroanisole, 3-bromo-4-fluorotrifluoromethoxybenzene, pubchem12363, acmc-1cocq
2-bromo-1-fluoro-4-(trifluoromethoxy)benzene

Specifications

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene
286932-57-8
MFCD02181186
VVVFPEJORKPEST-UHFFFAOYSA-N
2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
25067359
98%
5g
C7H3BrF4O
2-bromo-1-fluoro-4-trifluoromethoxy benzene, 1-bromo-2-fluoro-5-trifluoromethoxy benzene, 3-bromo-4-fluoro trifluoromethoxy benzene, 3-bromo-4-fluorotrifluoroanisole, 2-fluoro-5-trifluoromethoxy-bromobenzene, 3-bromo-4-fluorophenyl trifluoromethyl ether, 3-bromo-4-fluoro-alpha,alpha,alpha-trifluoroanisole, 3-bromo-4-fluorotrifluoromethoxybenzene, pubchem12363, acmc-1cocq
C1=CC(=C(C=C1OC(F)(F)F)Br)F
258.998
259
Videos
SDS Documents