Få mere at vide
(±)-1-Bromo-2-ethylhexane-d17, CDN
Artikelnummer. 30327908
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327908 0.5 g 0.5g
30325880 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327908 Leverandør CDN Leverandørnr. CDND74710.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-2-ethylhexane-D17
CAS 1398065-97-8
Molekylær formel C8 D17 Br
Mængde 0.5 g
Synonym (±)-1-Bromo-2-ethylhexane-d17, 3-(Bromomethyl-d2)heptane-1,1,1,2,2,3,4,4,5,5,6,6,7,7,7-d15, 1-Bromo-2-ethylhexane-D17, 1-Bromoiso-octane-d17, 2-Ethylhexyl-d17 bromide
InChI formel InChI=1 S/C8H17Br/c1-3-5-6-8(4-2)7-9/h8H,3-7H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])Br)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 3-[bromo(dideuterio)methyl]-1,1,1,2,2,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptane
Molekylvægt (g/mol) 210.2293
Formel vægt 209.1581 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity