Få mere at vide
1-Bromo-2,5-difluorobenzene-d3, CDN
Artikelnummer. 30324471
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324471 0.5 g 0.5g
30316826 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324471 Leverandør CDN Leverandørnr. CDND64730.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-2,5-difluorobenzene-D3
CAS 1219795-54-6
Molekylær formel C6 D3 Br F2
Mængde 0.5 g
Synonym Benzene-1,2,4-d3, 5-bromo-3,6-difluoro- (ACI), 5-Bromo-3,6-difluorobenzene-1,2,4-d3, 2-Bromo-1,4-difluorobenzene-d3, 1-Bromo-2,5-difluorobenzene-d3, 1-Bromo-2,5-difluorobenzene-D3
InChI formel InChI=1 S/C6H3BrF2/c7-5-3-4(8)1-2-6(5)9/h1-3 H/i1D,2 D,3 D
SMIL [2 H]c1c([2 H])c(F)c(Br)c([2 H])c1F
IUPAC navn 1-bromo-2,4,5-trideuterio-3,6-difluorobenzene
Molekylvægt (g/mol) 196.0073
Formel vægt 194.9574 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity