missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Bromo-2,3-dichlorobenzene, TRC

Artikelnummer. 30340889
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340889 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30340889 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6874351G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Bromo-2,3-dichlorobenzene
CAS 56961-77-4
Molekylær formel C6H3BrCl2
Mængde 1 g
Synonym 1-Bromo-2,3-dichloro-benzene, 2,3-Dichloro-1-bromobenzene, 2,3-Dichlorobromobenzene, 2,3-Dichlorophenyl Bromide
InChI formel InChI=1S/C6H3BrCl2/c7-4-2-1-3-5(8)6(4)9/h1-3H
SMIL Clc1cccc(Br)c1Cl
IUPAC navn 1-bromo-2,3-dichlorobenzene
Molekylvægt (g/mol) 225.9
Formel vægt 223.8795
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.