missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5,6-tetramethylbenzene, 99%, Thermo Scientific Chemicals

Catalog Number 11381277
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1646-53-3
C10H13Br
213.118
MFCD00000070
WJKBPTLQJXKEHC-UHFFFAOYSA-N
bromodurene, 3-bromodurene, 1-bromo-2,3,5,6-tetramethylbenzene, benzene, 3-bromo-1,2,4,5-tetramethyl, bromdurol, duryl bromide, bromodurene, pract., acmc-1bz04, 2,3,5,6-tetramethylbromobenzene, 1-bromo-2,5,6-tetramethylbenzene
74240
3-bromo-1,2,4,5-tetramethylbenzene
CC1=CC(=C(C(=C1C)Br)C)C
This item is not returnable. View return policy
1646-53-3
C10H13Br
213.118
MFCD00000070
WJKBPTLQJXKEHC-UHFFFAOYSA-N
bromodurene, 3-bromodurene, 1-bromo-2,3,5,6-tetramethylbenzene, benzene, 3-bromo-1,2,4,5-tetramethyl, bromdurol, duryl bromide, bromodurene, pract., acmc-1bz04, 2,3,5,6-tetramethylbromobenzene, 1-bromo-2,5,6-tetramethylbenzene
74240
3-bromo-1,2,4,5-tetramethylbenzene
CC1=CC(=C(C(=C1C)Br)C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Boiling Point 115°C to 120°C (5 mmHg)
Quantity 1 g
Beilstein 2207158
Formula Weight 213.12
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetramethylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'