Learn More
Sartorius 1 Branch Microsart™ Manifolds
Filtration stand designed to adapt to a variety of different needs.
Brand: Sartorius 168M1-BS250
Additional Details : Weight : 1.14000kg
Description
- 100% stainless steel for minimized cleaning effort
- Full autoclavability without disassembly
- Sterile venting in critical applications
- Reduced risk of secondary contamination
- Quick connection adapters for fast connection of tubing and fast interconnection of 2 manifolds
- Low working height for ergonomic working in laminar flow
- Accurately detects and quantifies microorganisms in liquid samples
- Method of choice for liquid testing
- Provides reliable and reproducible results
Specifications
Biosart 250 Funnels | |
1 | |
176 mm | |
120 mm |
Stainless Steel | |
250 mL | |
98 mm |