missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indoline-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7055333
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
143262-20-8
C14H17NO4
263.293
MFCD04973983
SUAMIYWLXFROHE-UHFFFAOYSA-N
n-boc-indoline-7-carboxylic acid, 1-tert-butoxycarbonyl indoline-7-carboxylic acid, 2,3-dihydro-indole-1,7-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-2,3-dihydroindole-7-carboxylic acid, n-boc-indoline-7-carboxylicacid, acmc-1c2ys, 1-tert-butoxycarbonyl-7-indolinecarboxylic acid, 1h-indole-1,7-dicarboxylic acid,2,3-dihydro-,1-1,1-dimethylethyl ester, 11bs-2,6-bis 3,5-bis trifluoromethyl phenyl-4-hydroxy-dinaphtho 2,1-d:1 inverted exclamation marka,2 inverted exclamation marka-f-1,3,2-dioxaphosphepin 4-oxide
14977845
1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-7-carboxylic acid
CC(C)(C)OC(=O)N1CCC2=CC=CC(=C21)C(=O)O
This item is not returnable. View return policy
143262-20-8
C14H17NO4
263.293
MFCD04973983
SUAMIYWLXFROHE-UHFFFAOYSA-N
n-boc-indoline-7-carboxylic acid, 1-tert-butoxycarbonyl indoline-7-carboxylic acid, 2,3-dihydro-indole-1,7-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-2,3-dihydroindole-7-carboxylic acid, n-boc-indoline-7-carboxylicacid, acmc-1c2ys, 1-tert-butoxycarbonyl-7-indolinecarboxylic acid, 1h-indole-1,7-dicarboxylic acid,2,3-dihydro-,1-1,1-dimethylethyl ester, 11bs-2,6-bis 3,5-bis trifluoromethyl phenyl-4-hydroxy-dinaphtho 2,1-d:1 inverted exclamation marka,2 inverted exclamation marka-f-1,3,2-dioxaphosphepin 4-oxide
14977845
1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-7-carboxylic acid
CC(C)(C)OC(=O)N1CCC2=CC=CC(=C21)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 182°C (decomposition)
Color Yellow
Quantity 250 mg
UN Number UN3077
Sensitivity Light sensitive
Formula Weight 263.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-indoline-7-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'