Learn More
1-(Boc-amino)cyclopentanecarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog Number 11388826
Click to view available options
Quantity:
1 g
5 g
35264-09-6
C11H19NO4
229.276
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
35264-09-6
C11H19NO4
229.276
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 134°C
Beilstein 2734412
Formula Weight 229.28
Percent Purity 98%
Quantity 5 g
Chemical Name or Material 1-(Boc-amino)cyclopentanecarboxylic acid