Få mere at vide
1-Boc-4-(6-aminopyridin-3-yl)piperazine-d4, TRC
Artikelnummer. 30329657
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30329657 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329657 Leverandør Toronto Research Chemicals Leverandørnr. TRCB62116250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Boc-4-(6-aminopyridin-3-yl)piperazine-d4
CAS 1808106-94-6
Molekylær formel C4 H8 O5 S
Mængde 50 mg
Synonym 4-(6-Aminopyridin-3-yl)piperazine-1-carboxylic Acid tert-Butyl Ester-d4, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate-d4, tert-Butyl 4-(6-Aminopyridin-3-yl)piperazine-1-carboxylate-d4
InChI formel InChI=1S/C4H8O5S/c1-3(4(5)6)9-10(2,7)8/h3H,1-2H3,(H,5,6)/t3-/m0/s1
SMIL C[C@H](OS(=O)(=O)C)C(=O)O
IUPAC navn (2S)-2-methylsulfonyloxypropanoic acid
Molekylvægt (g/mol) 168.168
Formel vægt 168.009