Få mere at vide
1-BOC-2,3-Dihydropyrrole-4-boronic acid, pinacol ester, TRC
Artikelnummer. 30342628
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30342628 100 mg
30340703 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30342628 Leverandør Toronto Research Chemicals Leverandørnr. TRCB201878100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-BOC-2,3-Dihydropyrrole-4-boronic acid, pinacol ester
CAS 1401165-14-7
Molekylær formel C15 H26 B N O4
Mængde 100 mg
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrole-1-carboxylic Acid tert-Butyl Ester
InChI formel InChI=1S/C15H26BNO4/c1-13(2,3)19-12(18)17-9-8-11(10-17)16-20-14(4,5)15(6,7)21-16/h10H,8-9H2,1-7H3
SMIL CC(C)(C)OC(=O)N1CCC(=C1)B2OC(C)(C)C(C)(C)O2
IUPAC navn tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydropyrrole-1-carboxylate
Molekylvægt (g/mol) 295.182
Formel vægt 295.195