Få mere at vide
1-(Benzyloxy)-4-bromo-2-methoxybenzene, TRC
Artikelnummer. 30342530
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30342530 250 mg
30331710 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30342530 Leverandør Toronto Research Chemicals Leverandørnr. TRCB291493250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(Benzyloxy)-4-bromo-2-methoxybenzene
CAS 63057-72-7
Molekylær formel C14H13BrO2
Mængde 250 mg
InChI formel InChI=1S/C14H13BrO2/c1-16-14-9-12(15)7-8-13(14)17-10-11-5-3-2-4-6-11/h2-9H,10H2,1H3
SMIL COc1cc(Br)ccc1OCc2ccccc2
IUPAC navn 4-bromo-2-methoxy-1-phenylmethoxybenzene
Molekylvægt (g/mol) 293.16
Formel vægt 292.0099