missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Benzyloxy-2-propanol, TRC

Artikelnummer. 30345718
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345718 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30345718 Leverandør Toronto Research Chemicals Leverandørnr. TRCB287755100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Benzyloxy-2-propanol
CAS 13807-91-5
Molekylær formel C10 H14 O2
Mængde 100 mg
Synonym 3-(Benzyloxy)-2-propanol, NSC 165612
InChI formel InChI=1S/C10H14O2/c1-9(11)7-12-8-10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3
SMIL OC(C)COCC=1C=CC=CC1
IUPAC navn 1-benzyloxypropan-2-ol
Molekylvægt (g/mol) 166.22
Formel vægt 166.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.