Få mere at vide
1-Benzyl-5-(1-(2,3-dimethylphenyl)vinyl)-1H-imidazole, TRC
Artikelnummer. 30135768
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135768 10 mg
30005147 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135768 Leverandør Toronto Research Chemicals Leverandørnr. TRCB25140010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2250243-56-0
Molekylær formel C20H20N2
Molekylvægt (g/mol) 288.39
Synonym N-Benzyl Vinyl Analog Medetomidine
IUPAC navn 1-benzyl-5-[1-(2,3-dimethylphenyl)ethenyl]imidazole
SMIL Cc1cccc(C(=C)c2cncn2Cc3ccccc3)c1C
Tekniske data
Kemisk navn eller materiale 1-Benzyl-5-(1-(2,3-dimethylphenyl)vinyl)-1H-imidazole
Analyt- eller komponentnavne 1-Benzyl-5-(1-(2,3-dimethylphenyl)vinyl)-1H-imidazole
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C20H20N2/c1-15-8-7-11-19(16(15)2)17(3)20-12-21-14-22(20)13-18-9-5-4-6-10-18/h4-12,14H,3,13H2,1-2H3
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 10 mg
Formel vægt 288.1626