missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Benzyl-4-[(5-benzyloxy-6-methoxy-1-indanone)-2-ylidenyl]methylpiperidine, TRC

Artikelnummer. 30335005
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30335005 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30335005 Leverandør Toronto Research Chemicals Leverandørnr. TRCB23295025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-Benzyl-4-[(5-benzyloxy-6-methoxy-1-indanone)-2-ylidenyl]me
CAS 120013-75-4
Molekylær formel C30 H31 N O3
Mængde 25 mg
Synonym 1H-Inden-1-one, 2,3-dihydro-6-methoxy-5-(phenylmethoxy)-2-[[1-(phenylmethyl)-4-piperidinyl]methylene]-, 1-Benzyl-4-[(5-benzyloxy-6-methoxy-1-indanone)-2-ylidenyl]methylpiperidine
InChI formel InChI=1S/C30H31NO3/c1-33-28-19-27-25(18-29(28)34-21-24-10-6-3-7-11-24)17-26(30(27)32)16-22-12-14-31(15-13-22)20-23-8-4-2-5-9-23/h2-11,16,18-19,22H,12-15,17,20-21H2,1H3
SMIL COc1cc2C(=O)C(=CC3CCN(Cc4ccccc4)CC3)Cc2cc1OCc5ccccc5
IUPAC navn 2-[(1-benzylpiperidin-4-yl)methylidene]-6-methoxy-5-phenylmethoxy-3H-inden-1-one
Molekylvægt (g/mol) 453.57
Formel vægt 453.2304
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.