Få mere at vide
1-Benzyl-3-hydroxy-4,5-bis(hydroxymethyl)-2-methylpyridin-1-ium-d2 Bromide, TRC
Artikelnummer. 30330238
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30330238 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330238 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27993325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Benzyl-3-hydroxy-4,5-bis(hydroxymethyl)-2-methylpyridin-1-ium-d2 Bromide
CAS 82896-37-5
Molekylær formel C15H16D2BrNO3
Mængde 25 mg
InChI formel InChI=1S/C15H17NO3.BrH/c1-11-15(19)14(10-18)13(9-17)8-16(11)7-12-5-3-2-4-6-12;/h2-6,8,17-18H,7,9-10H2,1H3;1H/i9D2;
SMIL [Br-].[2H]C([2H])(O)c1c[n+](Cc2ccccc2)c(C)c(O)c1CO
IUPAC navn 1-benzyl-5-[dideuterio(hydroxy)methyl]-4-(hydroxymethyl)-2-methylpyridin-1-ium-3-ol;bromide
Molekylvægt (g/mol) 342.22
Formel vægt 341.0596