Få mere at vide
1-Benzyl-1H-indazol-3-ol, Mikromol™
Artikelnummer. 30049407
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049407 25 mg
16314396 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049407 Leverandør Mikromol Leverandørnr. MM0142.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Benzyl-1H-indazol-3-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Benzyl-1H-Indazol-3-Ol
CAS 2215-63-6
Analyt- eller komponentnavne 1-Benzyl-1H-Indazol-3-Ol
Forsendelsestilstand Room Temperature
Molekylær formel C14 H12 N2 O
Certificeringer/compliance ISO 9001
Synonym 1,2-Dihydro-1-(phenylmethyl)-3H-indazol-3-one, 1-Benzyl-1,2-dihydro-3H-indazol-3-one, 1-Benzyl-1H-indazol-3-ol, 3-Hydroxy-1-benzyl-1H-indazole, 1-Benzyl-3-indazolone, NSC 247064, Benzydamine Hydrochloride Imp. C (BP), Benzydamine Hydrochloride Impurity C, Benzydamine Impurity C
InChI formel InChI=1S/C14H12N2O/c17-14-12-8-4-5-9-13(12)16(15-14)10-11-6-2-1-3-7-11/h1-9H,10H2,(H,15,17)
SMIL Oc1nn(Cc2ccccc2)c3ccccc13
IUPAC navn 1-benzylindazol-3-ol
Molekylvægt (g/mol) 224.26
Mængde 25 mg
Formel vægt 224.095 g/mol