Få mere at vide
1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride, Mikromol™
Artikelnummer. 30050998
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050998 25 mg
30052522 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050998 Leverandør Mikromol Leverandørnr. MM0362.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride
CAS 2731998-18-6
Analyt- eller komponentnavne 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N3 O . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym Azelastine Hydrochloride Imp. B (EP), Azelastine Imp. B (EP), Azelastine USP Related Compound B, Azelastine USP RC B, 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Dihydrochloride, Azelastine Hydrochloride Imp. B (EP) as Dihydrochloride, Azelastine Related Compound B as Dihydrochloride, Azelastine Hydrochloride Impurity B as Dihydrochloride, Azelastine Impurity B as Dihydrochloride
InChI formel InChI=1S/C14H21N3O.2ClH/c1-17-10-5-8-13(9-11-17)15-16-14(18)12-6-3-2-4-7-12;;/h2-4,6-7,13,15H,5,8-11H2,1H3,(H,16,18);2*1H
SMIL Cl.Cl.CN1CCCC(CC1)NNC(=O)c2ccccc2
IUPAC navn N'-(1-methylazepan-4-yl)benzohydrazide;dihydrochloride
Molekylvægt (g/mol) 320.26
Mængde 25 mg
Formel vægt 319.1218 g/mol