Learn More
1-Benzothiophene-3-carboxylic acid, 97%, Thermo Scientific™
Catalog Number p-2695553
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
5381-25-9
C9H6O2S
178.205
MFCD01846406
DRBLTQNCQJXSNU-UHFFFAOYSA-N
benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
601280
1-benzothiophene-3-carboxylic acid
C1=CC=C2C(=C1)C(=CS2)C(=O)O
5381-25-9
C9H6O2S
178.205
MFCD01846406
DRBLTQNCQJXSNU-UHFFFAOYSA-N
benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
601280
1-benzothiophene-3-carboxylic acid
C1=CC=C2C(=C1)C(=CS2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carboxylic acid