Learn More
1-Benzofuran-5-carbaldehyde, 97%, Thermo Scientific™
Product Code. 10661093
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661093 1 g Amber Glass Bottle 1g
10660903 250 mg Amber Glass Bottle 250mg
10001474 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10661093 Supplier Thermo Scientific Maybridge Supplier No. CC07104DA
 Restricted Item
Forventet forsendelse: 18-01-2027
Add to basket
Chemical Identifiers
CAS 10035-16-2
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD03411182
InChI Key LLLBDLDNTMMZHL-UHFFFAOYSA-N
Synonym benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde
PubChem CID 2773875
IUPAC Name 1-benzofuran-5-carbaldehyde
SMILES C1=CC2=C(C=CO2)C=C1C=O
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-carbaldehyde