Få mere at vide
1-Benzhydrylpiperazine-d8, TRC
Artikelnummer. 30334883
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30334883 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334883 Leverandør Toronto Research Chemicals Leverandørnr. TRCB196952100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Benzhydrylpiperazine-d8
CAS 1330188-81-2
Molekylær formel C17H12D8N2
Mængde 100 mg
Synonym 1-(Diphenylmethyl)piperazine-d8, 4-(Diphenylmethyl)piperazine-d8, 4-Benzhydrylpiperazine-d8, Benzhydrylpiperazine-d8, N-(Diphenylmethyl)piperazine-d8, N-Benzhydrylpiperazine-d8, NSC 35536-d8, Norcyclizine-d8
InChI formel InChI=1S/C17H20N2/c1-3-7-15(8-4-1)17(16-9-5-2-6-10-16)19-13-11-18-12-14-19/h1-10,17-18H,11-14H2/i11D2,12D2,13D2,14D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C(c2ccccc2)c3ccccc3)C1([2H])[2H]
IUPAC navn 1-benzhydryl-2,2,3,3,5,5,6,6-octadeuteriopiperazine
Molekylvægt (g/mol) 260.4
Formel vægt 260.2129