missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinol hydrochloride, 95%, Thermo Scientific™

Catalog Number p-7028480
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
90604-02-7
C16H18ClNO
275.78
MFCD00209447
LCHTWRWPHBRTNO-UHFFFAOYSA-N
1-benzhydrylazetidin-3-ol hydrochloride, 1-diphenylmethyl-3-hydroxyazetidine hydrochloride, 1-diphenylmethyl azetidin-3-ol hydrochloride, 1-benzhydryl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol-hcl, 1-benzhydrylazetidin-3-ol hcl, 1-benzhydryl azetidin-3-ol hcl, 1-diphenylmethyl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol hydrochloride, n-benzhydrylazetidin-3-ol hydrochloride
2801726
1-benzhydrylazetidin-3-ol;hydrochloride
Cl.OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
90604-02-7
C16H18ClNO
275.78
MFCD00209447
LCHTWRWPHBRTNO-UHFFFAOYSA-N
1-benzhydrylazetidin-3-ol hydrochloride, 1-diphenylmethyl-3-hydroxyazetidine hydrochloride, 1-diphenylmethyl azetidin-3-ol hydrochloride, 1-benzhydryl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol-hcl, 1-benzhydrylazetidin-3-ol hcl, 1-benzhydryl azetidin-3-ol hcl, 1-diphenylmethyl-3-azetidinol hydrochloride, 1-benzhydrylazetan-3-ol hydrochloride, n-benzhydrylazetidin-3-ol hydrochloride
2801726
1-benzhydrylazetidin-3-ol;hydrochloride
Cl.OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 174°C
Quantity 1 g
Formula Weight 275.78
Percent Purity 95%
Chemical Name or Material 1-Benzhydryl-3-azetidinol hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'